Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Glycolate 98.0+%, TCI America™
SDP

Catalog No. G0242100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0242100G 100 g
G024225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0242100G Supplier TCI America Supplier No. G0242100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-50-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00021970
InChI Key ZANNOFHADGWOLI-UHFFFAOYSA-N
Synonym ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
PubChem CID 12184
IUPAC Name ethyl 2-hydroxyacetate
SMILES CCOC(=O)CO

Specifications

Color Colorless
Boiling Point 160°C
Quantity 100 g
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Glycolate
RTECSNumber MC5410000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.