Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl iodoacetate

Catalog No. 24293425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-293-425G 25 g
24-293-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-293-425G Supplier Sigma Aldrich Supplier No. 24293425G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

Specifications

CAS 623-48-3
Density 1.808 g/mL (at 25°C (literature))
Boiling Point 179°C to 180°C (lit.)
Molecular Formula C4H7IO2
Linear Formula ICH2COOC2H5
Refractive Index n20/D 1.503 (literature)
MDL Number MFCD00001081
Quantity 25 g
Molecular Weight (g/mol) 214
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.