Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Isobutyl Ketone 98.0+%, TCI America™
SDP

Catalog No. E024910ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E024910ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E024910ML Supplier TCI America Supplier No. E024910ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 623-56-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00009319
InChI Key DXVYLFHTJZWTRF-UHFFFAOYSA-N
Synonym 5-Methyl-3-hexanone
PubChem CID 12187
IUPAC Name 5-methylhexan-3-one
SMILES CCC(=O)CC(C)C

Specifications

Color Yellow
Boiling Point 136°C
Quantity 10 mL
UN Number 1224
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isobutyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.