Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl isonipecotate

Catalog No. E3350525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3350525G 25 g
E33505100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E3350525G Supplier Sigma Aldrich Supplier No. E3350525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for synthesis of:
  • Smn protein modulators
  • α-aryl and α-amino-substituted aliphatic esters by rhodium catalyzed tandem double bond migration/conjugate addition
  • Nitroethylenediamines by nucleophilic ring opening of nitroimidazolidinone
  • Rhoa inhibitors for cardiovascular disease therapy
  • Saccharin derived mannich bases as antimicrobials and antioxidantsreactant for one-pot reductive amination and suzuki-miyaura cross coupling of formyl aryl and heteroaryl mida boronates

Specifications

CAS 1126-09-6
Density 1.02 g/mL (at 25°C (literature))
Boiling Point 204°C (lit.)
Molecular Formula C8H15NO2
Refractive Index n20/D 1.459 (literature)
Linear Formula C8H15NO2
MDL Number MFCD00006003
Quantity 25 g
Synonym Ethyl 4-piperidinecarboxylate; Piperidin-4-carboxylic acid ethyl ester
Molecular Weight (g/mol) 157.21
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.