Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isonipecotate, 98%

Catalog No. AAB2060122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2060122 100 g
AAB2060114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2060122 Supplier Thermo Scientific Chemicals Supplier No. B2060122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1126-09-6 | C8H15NO2 | 157.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is used as a reactant for synthesis of SMN protein modulators, β-aryl and β-amino-substituted aliphatic esters by rhodium catalyzed tandem double bond migration/conjugate addition, nitroethylenediamines by nucleophilic ring opening of nitroimidazolidinone. It is also involved in the reactions of RhoA inhibitors for cardiovascular disease therapy and saccharin derived Mannich bases as antimicrobials and antioxidants. It is employed as a reactant for one-pot reductive amination and Suzuki-Miyaura cross coupling of formyl aryl and heteroaryl MIDA boronates.

Solubility
Fully miscible with water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126-09-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00006003
InChI Key RUJPPJYDHHAEEK-UHFFFAOYSA-N
Synonym ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
PubChem CID 70770
IUPAC Name ethyl piperidine-4-carboxylate
SMILES CCOC(=O)C1CCNCC1

Specifications

Density 1.021
Boiling Point 203°C to 205°C
Flash Point 80°C (176°F)
Refractive Index 1.46
Quantity 100 g
Beilstein 118419
Solubility Information Fully miscible with water.
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material Ethyl isonipecotate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-416-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.