accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH

CAS RN 1126-09-6

IUPAC Name: ethyl piperidine-4-carboxylate
Synonyms: ethyl piperidine-4-carboxylate
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: RUJPPJYDHHAEEK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCNCC1
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (4)
  • (1)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)