Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl L-Lactate, 98%, Spectrum™ Chemical
SDP

Catalog No. 18610432 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
98%
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-432 100 mL Amber Glass Bottle
18-610-436 500 mL 98%
18-610-435 4 L 98%
18-610-433 20 L 98%
18-610-434 200 L 98%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-610-432 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1187100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97-64-3

Specifications

Packaging Amber Glass Bottle
CAS Max % 1
Quantity 100 mL
Percent Purity 98%
Grade Reagent
Chemical Name or Material Ethyl L-Lactate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.