Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl L-(+)-Mandelate 98.0+%, TCI America™
SDP

Catalog No. M127325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M127325G 25 g
M12735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M127325G Supplier TCI America Supplier No. M127325G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13704-09-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00064249
InChI Key SAXHIDRUJXPDOD-VIFPVBQESA-N
Synonym L-(+)-Mandelic Acid Ethyl Ester
PubChem CID 6951557
ChEBI CHEBI:78405
IUPAC Name ethyl (2S)-2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Color White-Yellow
Boiling Point 107°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl L-(+)-Mandelate
EINECSNumber (3)-3841
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.