Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl-m-Aminobenzoate, methanesulfonate salt, MP Biomedicals™

Catalog No. ICN10310605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10310605 5 g
ICN10310625 25 g
ICN10310680 100 g
ICN10310683 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10310605 Supplier MP Biomedicals, Inc Supplier No. 0210310605
Add to Cart
Add to Cart

Ethyl-m-Aminobenzoate is a derivative of an isomer of benzocaine

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-86-2
Molecular Formula C10H15NO5S
Molecular Weight (g/mol) 261.292
MDL Number MFCD00013176
InChI Key FQZJYWMRQDKBQN-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt
PubChem CID 261501
IUPAC Name ethyl 3-aminobenzoate;methanesulfonic acid
SMILES CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O

Specifications

Color White
Density Ethyl-m-Aminobenzoate is mainly used as an anesthetic in fish,sharks,frogs
Assay Percent Range 0.993
Quantity 5 g
Formula Weight 261.3
Physical Form Powder
Chemical Name or Material Ethyl-m-Aminobenzoate, Methanesulfonate Salt
EINECSNumber 212-956-8
Recommended Storage Store at -0°C. Keep away from sunlight. Sunlight will cause degradation.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.