Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl malonyl chloride

Catalog No. 1638725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-387-25G 5 g
16-387-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-387-25G Supplier Sigma Aldrich Supplier No. 1638725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl malonyl chloride is a versatile acylating agent for propargyl alcohols, hydrazines and amines.ethyl malonyl chloride was used in the synthesis of:
  • Liquid-crystalline methanofullerodendrimers
  • 3,5-disubstituted 1,2,4-oxadiazole derivatives, which are potential peptidomimetic building blocks
  • 3-pyrrolin-2-ones via amidation with propargylamines and subsequent base catalyzed 5-exo-dig cyclization

Specifications

CAS 36239-09-5
Density 1.176 g/mL (at 25°C (literature))
Boiling Point 79°C to 80°C (25 mmHg)
Molecular Formula C5H7ClO3
Refractive Index n20/D 1.429 (literature)
Linear Formula CH3CH2OCOCH2COCl
MDL Number MFCD00000736
Quantity 5 g
Synonym Ethyl (chloroformyl)acetate; Ethyl 3-chloro-3-oxopropionate
Molecular Weight (g/mol) 150.56
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.