Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl N-BOC-4-piperidinecarboxylate, 97%

Catalog No. AC431500050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431500050 5 g Glass bottle
AC431500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431500050 Supplier Thermo Scientific Chemicals Supplier No. 431500050
Add to Cart

CAS: 142851-03-4 | C13H23NO4 | 257.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142851-03-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD01763998
InChI Key MYHJCTUTPIKNAT-UHFFFAOYSA-N
Synonym ethyl n-boc-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate, ethyl n-boc-4-piperidinecarboxylate, n-boc-4-carbethoxy piperidine, 1,4-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 4-ethyl ester, ethyl n-boc-isonipecotate, ethyl 1-boc-piperidine-4-carboxylate, dimethylethyl piperidinedicarboxylic acid ethyl ester, 1-boc-piperidine-4-carboxylic acid ethyl ester, 1-boc-isonipecotic acid ethyl ester
PubChem CID 2758812
IUPAC Name 1-O-tert-butyl 4-O-ethyl piperidine-1,4-dicarboxylate
SMILES CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Color Colorless to Yellow
Density 1.0460g/mL
Boiling Point 120.0°C to 135.0°C (0.5 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.046
Formula Weight 257.33
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl N-BOC-4-piperidinecarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.