Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Nicotinate 99.0+%, TCI America™
SDP

Catalog No. N0085100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0085100ML 100 mL
N008525ML 25 mL
N0085500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0085100ML Supplier TCI America Supplier No. N0085100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 614-18-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006389
InChI Key XBLVHTDFJBKJLG-UHFFFAOYSA-N
Synonym ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan
PubChem CID 69188
IUPAC Name ethyl pyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 224°C
Quantity 100 mL
Formula Weight 151.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Nicotinate
EINECSNumber (5)-6766
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.