Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl nitroacetate

Catalog No. 1923335G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-233-35G 5 g
19-233-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-233-35G Supplier Sigma Aldrich Supplier No. 1923335G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl nitroacetate has been used in:
  • Synthesis of α-oxoacids via michael addition reaction with α,α-unsaturated ketones
  • Fuctionalization of c4-position on pyrimidine and c6-position on 2α-deoxyguanosine to produce novel nucleosides
  • Facile synthesis of α,α-diisobutylglycine
  • Synthesis of dl-4,4-difluoroglutamic acid

Specifications

CAS 626-35-7
Density 1.199 g/mL (at 25°C (literature))
Boiling Point 105°C to 107°C (25 mmHg)
Molecular Formula C4H7NO4
Linear Formula NO2CH2CO2C2H5
Refractive Index n20/D 1.424 (literature)
MDL Number MFCD00007403
Quantity 5 g
Molecular Weight (g/mol) 133.1
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.