Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl oleate, 98%, mixture of homologeous fatty acid esters

Catalog No. p-4509554
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410292500 250 g Glass bottle
AC410290010 1 kg Glass bottle
AC410290025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC410292500 Supplier Thermo Scientific Chemicals Supplier No. 410292500
Only null left
Add to Cart

CAS: 111-62-6 | C20H38O2 | 310.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.51
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Acidity 0.5mg KOH/g max.
Density 0.8600g/mL
Boiling Point 190.0°C to 210.0°C (7.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 60% min. (oleic acid) (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4530
Linear Formula CH3(CH2)7CHCH(CH2)7COOC2H5
Quantity 250 g
Saponification Value 177 to 188mg KOH/g
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 310.51
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material Ethyl oleate
Show More Show Less
EINECSNumber 203-889-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.