Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Oleate, NF, Spectrum™ Chemical
SDP

Catalog No. 18602545 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-545 500 g Amber Glass Bottle
18-602-546 5 kg Poly Pail
18-602-543 25 g Amber Glass Bottle
18-602-544 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-545 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1172500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

E1172, 111-62-6, C20H38O2

Spectrum™ Chemical Ethyl Oleate, NF is used as a solubilizing agent in pharmaceutical preparations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.52
InChI Key LVGKNOAMLMIIKO-VAWYXSNFSA-N
IUPAC Name ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)OCC

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.