Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Pentafluoropropionate 98.0+%, TCI America™
SDP

Catalog No. P112325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P112325G 25 g
P11235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P112325G Supplier TCI America Supplier No. P112325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 426-65-3
Molecular Formula C5H5F5O2
Molecular Weight (g/mol) 192.085
MDL Number MFCD00000431
InChI Key DBOFMRQAMAZKQY-UHFFFAOYSA-N
Synonym ethyl pentafluoropropionate, ethyl perfluoropropionate, ethylpentafluoropropionate, ethyl pentafluoropropanoate, pentafluoropropionic acid ethyl ester, propanoic acid, pentafluoro-, ethyl ester, propanoic acid, 2,2,3,3,3-pentafluoro-, ethyl ester, 2,2,3,3,3-pentafluoro-propionic acid ethyl ester, pubchem12575, acmc-1asrx
PubChem CID 67928
IUPAC Name ethyl 2,2,3,3,3-pentafluoropropanoate
SMILES CCOC(=O)C(C(F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 76°C
Quantity 25 g
UN Number 3272
Formula Weight 192.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Pentafluoropropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.