Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenyl Sulfone, MP Biomedicals

Catalog No. ICN228457 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN228457 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN228457 Supplier MP Biomedicals, Inc Supplier No. 0522845705

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 599-70-2
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.226
InChI Key VBQUDDWATQWCPP-UHFFFAOYSA-N
Synonym ethyl phenyl sulfone, ethylsulfonyl benzene, ethylphenyl sulfone, ethyl phenyl sulphone, sulfone, ethyl phenyl, ethylsulphonyl benzene, ethylphenylsulfone, benzene, ethylsulfonyl, ethanesulfonyl benzene, 3blu
PubChem CID 69032
IUPAC Name ethylsulfonylbenzene
SMILES CCS(=O)(=O)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 170.23
Chemical Name or Material Ethyl Phenyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.