Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenyl Sulfone 97.0+%, TCI America™
SDP

Catalog No. E042725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E042725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E042725G Supplier TCI America Supplier No. E042725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 599-70-2
Melting Point 42°C
Color White-Yellow
Boiling Point 103°C
Molecular Formula C8H10O2S
Quantity 25 g
MDL Number MFCD00025081
Synonym ethyl phenyl sulfone, ethylsulfonyl benzene, ethylphenyl sulfone, ethyl phenyl sulphone, sulfone, ethyl phenyl, ethylsulphonyl benzene, ethylphenylsulfone, benzene, ethylsulfonyl, ethanesulfonyl benzene, 3blu
InChI Key VBQUDDWATQWCPP-UHFFFAOYSA-N
SMILES CCS(=O)(=O)C1=CC=CC=C1
IUPAC Name ethylsulfonylbenzene
Molecular Weight (g/mol) 170.226
PubChem CID 69032
Formula Weight 170.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Phenyl Sulfone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.