Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenylcyanoacetate 95.0+%, TCI America™
SDP

Catalog No. P125525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P125525G 25 g
P12555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P125525G Supplier TCI America Supplier No. P125525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4553-07-5
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001866
InChI Key SXIRJEDGTAKGKU-UHFFFAOYNA-N
Synonym ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
PubChem CID 95298
ChEBI CHEBI:51926
IUPAC Name ethyl 2-cyano-2-phenylacetate
SMILES CCOC(=O)C(C#N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 275°C
Quantity 25 g
Formula Weight 189.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Phenylcyanoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.