Learn More
Selleck Chemical LLC Pentadecanoic acid
SDP
Supplier:  Selleck Chemical LLC S623425MG
Encompass_Preferred
Pentadecanoic acid is a saturated fatty acid of exogenous (primarily ruminant) origin It also has a role as a plant metabolite a food component a Daphnia magna metabolite a human blood serum metabolite and an algal metabolite
Catalog No. 50-000-03278
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.