Learn More
Apexbio Technology LLC VU 0364439 1246086-78-1 25mg
SDP
Supplier:  Apexbio Technology LLC A262325
Encompass_Preferred
VU 0364439 (CAS 1246086-78-1) is a positive allosteric modulator (PAM) of the metabotropic glutamate receptor 4 (mGluR4) a member of the class C G protein-coupled receptor (GPCR) family involved in regulating glutamatergic neurotransmission In vitro studies have demonstrated that VU 0364439 enhances mGluR4 activity with an EC50 of 19 8 nM exhibiting greater maximal response and potency compared to the partially selective PAM ()-PHCCC Although its pharmacokinetic properties limit in vivo applications VU 0364439 serves as a valuable tool compound for investigating mGluR4 function and modulation in cellular assays
Catalog No. 50-000-93375
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.