Learn More
Medchemexpress LLC Ethyl pyruvate | 617-35-6 | MFCD00009123 | 250mg
SDP
Supplier:  Medchemexpress LLC HYY1362250MG
Encompass_Preferred
Ethyl pyruvate is a simple derivative of the endogenous metabolite pyruvate Ethyl pyruvate is an HMGB1 release inhibitor Ethyl pyruvate can induce apoptosis by autophagy Ethyl pyruvate has anti-inflammatory antioxidant and anti-tumor activity Ethyl pyruvate can be used in the study of neurodegenerative diseases such as Alzheimer s and Parkinson s disease[1][2][3][4][5]
Catalog No. 50-002-67794
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.