Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%, Thermo Scientific™

Catalog No. AAH6150714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6150714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6150714 Supplier Thermo Scientific Chemicals Supplier No. H6150714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90315-82-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00077794
InChI Key ZJYKSSGYDPNKQS-LLVKDONJSA-N
Synonym ethyl r-2-hydroxy-4-phenylbutyrate, r-ethyl 2-hydroxy-4-phenylbutanoate, ethyl 2r-2-hydroxy-4-phenylbutanoate, ethyl r---2-hydroxy-4-phenylbutyrate, r---2-hydroxy-4-phenylbutyrate ethyl ester, qp7iwh2qox, unii-qp7iwh2qox, r-2-hydroxy-4-phenylbutyric acid ethyl ester, r-hpbe, r-2-hydroxy-4-phenyl butyric acid ethyl ester
PubChem CID 2733848
IUPAC Name ethyl (2R)-2-hydroxy-4-phenylbutanoate
SMILES CCOC(=O)C(CCC1=CC=CC=C1)O

Specifications

Color Colorless
Density 1.075 g/mL
Boiling Point 212°C
Flash Point 150°C (302°F)
Refractive Index 1.504
Quantity 25 g
Beilstein 3590943
Formula Weight 208.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl (R)-2-hydroxy-4-phenylbutyrate
Show More Show Less
EINECSNumber 618-525-4
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.