accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90315-82-5

IUPAC Name: ethyl (2R)-2-hydroxy-4-phenylbutanoate
Synonyms: ethyl (2R)-2-hydroxy-4-phenylbutanoate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: ZJYKSSGYDPNKQS-LLVKDONJSA-N
SMILES: CCOC(=O)[C@H](O)CCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)