Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-2-Hydroxy-4-phenylbutyrate 98.0+%, TCI America™
SDP

Catalog No. H09045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09045G Supplier TCI America Supplier No. H09045G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90315-82-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00077794
InChI Key ZJYKSSGYDPNKQS-LLVKDONJSA-N
Synonym ethyl r-2-hydroxy-4-phenylbutyrate, r-ethyl 2-hydroxy-4-phenylbutanoate, ethyl 2r-2-hydroxy-4-phenylbutanoate, ethyl r---2-hydroxy-4-phenylbutyrate, r---2-hydroxy-4-phenylbutyrate ethyl ester, qp7iwh2qox, unii-qp7iwh2qox, r-2-hydroxy-4-phenylbutyric acid ethyl ester, r-hpbe, r-2-hydroxy-4-phenyl butyric acid ethyl ester
PubChem CID 2733848
IUPAC Name ethyl (2R)-2-hydroxy-4-phenylbutanoate
SMILES CCOC(=O)C(CCC1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-2-Hydroxy-4-phenylbutyrate
EINECSNumber (3)-4129
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.