Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-(-)-4-cyano-3-hydroxybutyrate, 98%

Catalog No. AAL1977114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1977114 25 g
AAL1977106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1977114 Supplier Thermo Scientific Chemicals Supplier No. L1977114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 141942-85-0 | C7H11NO3 | 157.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141942-85-0
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00270839
InChI Key LOQFROBMBSKWQY-ZCFIWIBFSA-N
Synonym ethyl r---4-cyano-3-hydroxybutyate, ethyl r---4-cyano-3-hydroxybutyrate, ethyl 3r-4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxybutyric acid ethyl ester, r-ethyl 4-cyano-3-hydroxybutyrate, ethyl-r-+-4-cyano-3-hydroxybutanoate, ethyl r-4-cyano-3-hydroxybutyrate, r-ethyl 4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxy-butyric acid ethyl ester, r---4-cyano-3-hydroxybutyric acid ethyl ester
PubChem CID 2733879
IUPAC Name ethyl (3R)-4-cyano-3-hydroxybutanoate
SMILES CCOC(=O)C[C@H](O)CC#N

Specifications

Density 1.114
Boiling Point 270°C
Flash Point >110°C (230°F)
Refractive Index 1.448
Quantity 25 g
Beilstein 5328715
Optical Rotation −4.5° (Neat)
Formula Weight 157.17
Percent Purity 98%
Chemical Name or Material Ethyl (R)-(-)-4-cyano-3-hydroxybutyrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 430-220-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.