Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Red 97.0+%, TCI America™
SDP

Catalog No. D05241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05241G 1 g
D052425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05241G Supplier TCI America Supplier No. D05241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76058-33-8
Molecular Formula C17H18N3O2
Molecular Weight (g/mol) 296.35
MDL Number MFCD00002427
InChI Key HBRCDTRQDHMTDA-UHFFFAOYSA-M
PubChem CID 54832
IUPAC Name 2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
SMILES CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 146°C
Synonym 4-(Diethylamino)azobenzene-2′C-carboxylic Acid, Diethyl Red
Formula Weight 297.36
Percent Purity ≥97.0% (T)
Color Purple/Red
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Ethyl Red [not Cyanin dyes]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.