Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (S)-(+)-3-Hydroxybutyrate 96.0+%, TCI America™
SDP

Catalog No. H097525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H097525G 25 g
H09755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H097525G Supplier TCI America Supplier No. H097525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
PubChem CID 6950306
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (S)-(+)-3-Hydroxybutyrate
EINECSNumber (2)-1430
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.