Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (S)-3-Piperidinecarboxylate D-Tartrate 98.0+%, TCI America™
SDP

Catalog No. N068125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N068125G 25 g
N06815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N068125G Supplier TCI America Supplier No. N068125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83602-38-4
Molecular Formula C12H21NO8
Molecular Weight (g/mol) 307.299
MDL Number MFCD03093639
InChI Key HHPGQKZOPPDLNH-FWQMQHHGSA-N
Synonym (S)-Nipecotic Acid Ethyl Ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, (S)-3-Piperidinecarboxylic Acid Ethyl Ester D-Tartrate
PubChem CID 118855422
IUPAC Name (3S)-2,3-dihydroxybutanedioic acid;ethyl piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 156°C
Color White
Quantity 25 g
Formula Weight 307.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (S)-3-Piperidinecarboxylate D-Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.