Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Tridecanoate 97.0+%, TCI America™
SDP

Catalog No. T095925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T095925ML 25 mL
T09595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T095925ML Supplier TCI America Supplier No. T095925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28267-29-0
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00008978
InChI Key QJYYMNOTJXIOBP-UHFFFAOYSA-N
Synonym tridecanoic acid ethyl ester, tridecanoic acid, ethyl ester, ethyltridecanoate, ethyl=tridecanoate, tridecanoic acid, ethylester, acmc-1cr51, ethyl tridecanoate, n-tridecanoic acid ethyl ester
PubChem CID 119908
IUPAC Name ethyl tridecanoate
SMILES CCCCCCCCCCCCC(=O)OCC

Specifications

Color Colorless
Boiling Point 198°C
Quantity 25 mL
Formula Weight 242.40
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Tridecanoate
EINECSNumber (2)-0798
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.