Learn More
Ethyl trifluoroacetoacetate, 97%
Description
Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.
Solubility
Miscible with water, ethanol, benzene and organic solvents.
Notes
Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications
Specifications
| Melting Point | -30°C |
| Density | 1.259 |
| Boiling Point | 129°C to 130°C |
| Flash Point | 38°C (100°F) |
| Odor | Odorless |
| Refractive Index | 1.376 |
| Quantity | 50 g |
| UN Number | UN3272 |
| Beilstein | 608353 |
| Solubility Information | Miscible with water,ethanol,benzene and organic solvents. |
| Show More |
Safety and Handling
| Hazard Category | H226-H302 |
| Hazard Statement | GHS H Statement H226-H302 Flammable liquid and vapour. Harmful if swallowed. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S. |
| EINECSNumber | 206-750-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.