Learn More
Sigma Organic Chemistry Ethyl vinyl ether | 1L | 109-92-2 | MFCD00009248 | 0.99
Supplier: Sigma Organic Chemistry 4221771L

Ethyl vinyl ether, 1L
About This Item:
Synonym(s): Ethoxyethylene
Linear Formula: C2H5OCH=CH2
Molecular Weight: 72.11
CAS Number: 109-92-2
MDL Number: MFCD00009248
UNSPSC Code: 12162002
Purity: 0.99
Boiling Point: 33 C (lit.)
Melting Point: -116 C (lit.)
Density: 0.753 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.376 (lit.)
Storage: 2-8C
EINECS Number: 203-718-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.