Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Violet, 0.1% in 50% Methanol, Reagents

Catalog No. 01335393
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-393 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-393 Supplier Reagents Holdings Llc Supplier No. CE002500100C
Add to Cart
Add to Cart

Ethyl Violet, 0.1% in 50% Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Basic Violet 4, Ethyl purple 6B
IUPAC Name water
SMILES O

Specifications

Boiling Point 77°C
Percent Purity 0.1%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8973
Color Blue
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Ethyl Violet, 0.1% in 50% Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.