Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Violet

Catalog No. p-7042844
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6395114 25 g
AAJ6395122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6395114 Supplier Thermo Scientific Chemicals Supplier No. J6395114
Only null left
Add to Cart

CAS: 2390-59-2 | C31H42ClN3 | 492.15 g/mol

Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

Solubility
Soluble in water (10 mg/ml).

Notes
Dye content approx. 80%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-59-2
Molecular Formula C31H42ClN3
Molecular Weight (g/mol) 492.15
MDL Number MFCD00011765
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
Synonym C.I. 42600; Basic Violet 4
PubChem CID 16955
IUPAC Name [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Specifications

Beilstein 4117566
Solubility Information Soluble in water (10mg/ml).
Formula Weight 492.15
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Ethyl Violet
EINECSNumber 219-231-5
RTECSNumber KH2682000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.