Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene, ≥99 %

Catalog No. AAL059080F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL059080F 2500 mL
AAL05908AE 100 mL
AAL05908AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL059080F Supplier Thermo Scientific Chemicals Supplier No. L059080F
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 100-41-4 | C8H10 | 106.168 g/mol

Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.

Solubility
Miscible with alcohol, ethylalcohol, benzene, ethyl ether, carbon tetrachloride, organic solvents, sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Melting Point -95°C
Density 0.867
Boiling Point 135°C to 136°C
Flash Point 15°C (59°F)
Odor Petrol-like
Refractive Index 1.495
Quantity 2500 mL
UN Number UN1175
Beilstein 1901871
Merck Index 14,3765
Solubility Information Miscible with alcohol,ethylalcohol,benzene,ethyl ether,carbon tetrachloride,organic solvents,sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material Ethylbenzene
Show More Show Less
Hazard Category H225-H304-H332-H335-H336-H351-H373
Hazard Statement GHS H Statement
H225-H304-H373-H332
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P271-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLBENZENE
EINECSNumber 202-849-4
RTECSNumber DA0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.