CAS RN 100-41-4
Ethylbenzene, 99.8%, anhydrous, AcroSeal™
CAS: 100-41-4 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N Synonym: phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
Ethylbenzene, ≥99 %
CAS: 100-41-4 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00011647 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N Synonym: phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
Ethylbenzene, 99.8%, pure
CAS: 100-41-4 MDL Number: MFCD00011647 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N Synonym: phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
Ethylbenzene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards..
Ethylbenzene Solution, NMR Reference Standard, NMR Tube Size: 5 mm 8 in., MilliporeSigma™ Supelco™
0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01%
Ethylbenzene, Solstice
CAS: 100-41-4 PubChem CID: 7500 ChEBI: CHEBI:16101
Ethylbenzene, ≥99.5% (GC), MilliporeSigma™ Supelco™
Ethylbenzene is a colorless liquid, which has an aromatic odor. It is widely used in the manufacturing of styrene and as a solvent. It is insoluble in water.
Ethylbenzene 99.0+%, TCI America™
CAS: 100-41-4 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00011647 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N Synonym: phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
Ethylbenzene, SPEX CertiPrep™
CAS: 100-41-4 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
Agilent Technologies Ethylbenzene Solution, 100μg/mL in Methanol, Ultra Scientific
CAS: 100-41-4 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
Ethylbenzene (Certified), Fisher Chemical
CAS: 100-41-4 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00011647 InChI Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N Synonym: phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene PubChem CID: 7500 ChEBI: CHEBI:16101 IUPAC Name: ethylbenzene SMILES: CCC1=CC=CC=C1
- 1
- 2