Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene, EMD-Millipore

Catalog No. M1096350005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096350005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096350005 Supplier MilliporeSigma Supplier No. 1096350005

Reference substance for gas chromatography

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Packaging Glass
Explosion Limit 1.0 to 7.8 %(V)
Ignition Point 430°C
Vapor Pressure 9.5 hPa (20°C)
Density Density: 0.87g/cm3 (20°C)
Quantity 5 mL
Formula Weight 106.16
Chemical Name or Material Ethylbenzene
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.