Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene, Solstice

Catalog No. 60090003 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-003 250 mL
60-090-002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-003 Supplier Solstice Supplier No. 03080250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ≥99.0% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-41-4
PubChem CID 7500
ChEBI CHEBI:16101

Specifications

Boiling Point 135°C to 137°C (1.013 hPa)
Flash Point 15 °C
Packaging Glass Bottle
Refractive Index 1.494 - 1.496
Quantity 250 mL
Percent Purity min. 99.0 %
Grade ACS Grade Plus
Physical Form Liquid
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.