Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene/ethyl Acrylate Copolymer, Thermo Scientific™

Catalog No. AC190891000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190891000 100 g
AC190895000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190891000 Supplier Thermo Scientific Chemicals Supplier No. 190891000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9010-86-0
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
InChI Key CGPRUXZTHGTMKW-UHFFFAOYSA-N
Synonym ethene; ethyl prop-2-enoate, poly ethylene-ethyl acrylate, 2-propenoic acid, ethyl ester, polymer with ethene, ethylene/ethyl acrylate, ethyl acrylate; ethylene
PubChem CID 3080657
IUPAC Name ethene;ethyl prop-2-enoate
SMILES CCOC(=O)C=C.C=C

Specifications

Quantity 100 g
Chemical Name or Material Ethylene/ethyl acrylate copolymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.