Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol AR, Macron Fine Chemicals™

Catalog No. MK50011 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
5 gL
4 x 4 L
Packaging:
Glass
Poly
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK5001-1 1 L Glass
MK-5001-5 5 gL Poly Pail
MK-5001-4 4 x 4 L Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK5001-1 Supplier Avantor Supplier No. 500106

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Quantity 1 L
Packaging Glass
Formula Weight 62.07
Chemical Name or Material Ethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.