Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol butyl ether, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019159 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9159 5 mL
11-102-0588 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9159 Supplier MilliporeSigma Supelco Supplier No. 530715MLF
Add to Cart
Add to Cart

Ethylene glycol butyl ether can find applications as a surfactant in investigating the process of spreading, shrinking and resting regime on the surface of water. It can also find applications as a cosolvent in water-borne paints.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 473°F
CAS 111-76-2
Vapor Pressure 10 mmHg (81°C)
Vapor Density 4.1 (vs. air)
Density 0.902 g/mL (at 25°C)
Flash Point 67°C
Physical Form Neat
Linear Formula CH3(CH2)3 OCH2 CH2 OH
MDL Number MFCD00002884
Melting Point -75°C
Boiling Point 169°C to 172.5°C
Synonym 2-Butoxyethanol; Butyl glycol
Type Analytical Standard
Quantity 5 mL
Formula Weight 118.17
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.