Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Dibenzyl Ether 95.0+%, TCI America™
SDP

Catalog No. D016025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D016025ML 25 mL
D01605ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D016025ML Supplier TCI America Supplier No. D016025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 622-22-0
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.32
MDL Number MFCD00030016
InChI Key FPFHYKOBYMYVAN-UHFFFAOYSA-N
Synonym 1,2-Bis(benzyloxy)ethane, Dibenzyl Cellosolve, Dibenzyl Glycol, 1,2-Dibenzyloxyethane
PubChem CID 222542
IUPAC Name {[2-(benzyloxy)ethoxy]methyl}benzene
SMILES C(COCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 210°C
Quantity 25 mL
Formula Weight 242.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Dibenzyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.