Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Monovinyl Ether (stabilized with KOH) 98.0+%, TCI America™
SDP

Catalog No. E051825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E051825ML 25 mL
E0518500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E051825ML Supplier TCI America Supplier No. E051825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 764-48-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00192151
InChI Key VUIWJRYTWUGOOF-UHFFFAOYSA-N
Synonym Vinyl Cellosolve, Vinyl Glycol, 2-Vinyloxyethanol
PubChem CID 12995
IUPAC Name 2-ethenoxyethanol
SMILES C=COCCO

Specifications

Color Yellow
Boiling Point 143°C
Quantity 25 mL
UN Number 3271
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monovinyl Ether (stabilized with KOH)
EINECSNumber (7)-0097
RTECSNumber KM5495000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.