Learn More
Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications
Specifications
| CAS Max % | 100% |
| Acidity | 0.005% |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Boiling Point | 196°C to 199°C |
| Percent Purity | 99% |
| Grade | Reagent |
| Chemical Name or Material | Ethylene Glycol |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.