Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18602565 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-565 4 x 4 L Amber Glass Bottle
18-602-559 500 mL Poly Bottle
18-602-561 1 L Poly Bottle
18-602-560 6 x 500 mL Poly Bottle
18-602-562 4 L Amber Glass Bottle
18-602-563 4 L Poly Bottle
18-602-564 6 x 1 L Poly Bottle
18-602-566 4 x 4 L Poly Bottle
18-602-567 20 L Poly Drum
18-602-568 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 18-602-565 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E10514LTGLCS4
Add to Cart
Add to Cart

E1051, 107-21-1, C2H6O2

Spectrum™ Chemical Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

Acidity 0.005%
CAS Max % 100%
Quantity 4 x 4 L
Packaging Amber Glass Bottle
Boiling Point 196°C to 199°C
Percent Purity 99%
Grade Reagent
Chemical Name or Material Ethylene Glycol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.