Learn More
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | -13°C (literature) |
| CAS Max % | 100.0000% |
| Density | 1.113 g/mL at 25 °C(lit.) |
| Quantity | 30 kg |
| Flash Point | 111°C |
| Packaging | Stainless Steel Can |
| Linear Formula | HOCH2CH2OH |
| Beilstein | 505945 |
| Boiling Point | 197°C |
| Formula Weight | 62.07 |
| Show More |
Safety and Handling
| ShelfLife | 1080 days from date of manufacture |
| EINECSNumber | 203-473-3 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.