Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Trithiocarbonate 96.0+%, TCI America™
SDP

Catalog No. E0323100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0323100G 100 g
E032325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0323100G Supplier TCI America Supplier No. E0323100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 822-38-8
Molecular Formula C3H4S3
Molecular Weight (g/mol) 136.245
MDL Number MFCD00005493
InChI Key XCWPBWWTGHQKDR-UHFFFAOYSA-N
Synonym ethylene trithiocarbonate, cyclic ethylene trithiocarbonate, trithiocarbonic acid, cyclic ethylene ester, carbonic acid, trithio-, cyclic ethylene ester, cylic ethylene trithiocarbonate, acmc-209pnw, 1,3-dithiolan-2-thione, 5-19-04-00014 beilstein handbook reference, ksc449a8r
PubChem CID 13196
IUPAC Name 1,3-dithiolane-2-thione
SMILES C1CSC(=S)S1

Specifications

Quantity 100 g
Formula Weight 136.25
Percent Purity ≥96.0% (GC)
Physical Form Crystals
Chemical Name or Material Ethylene Trithiocarbonate
RTECSNumber FG2720000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.