Learn More
Sigma Organic Chemistry Ethylenediamine | 100ML | 107-15-3 | MFCD00008204
Supplier: Sigma Organic Chemistry E26266100ML
            Ethylenediamine, 100ML
About This Item:
Synonym(s): 1,2-Diaminoethane
Linear Formula: NH2CH2CH2NH2
Molecular Weight: 60.1
CAS Number: 107-15-3
MDL Number: MFCD00008204
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 118 C (lit.)
Melting Point: 8.5 C (lit.)
Density: 0.899 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.4565 (lit.)
Storage: room temp
EINECS Number: 203-468-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.