Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine Tetraacetic Acid, MP Biomedicals™

Catalog No. MP04800683 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04800683 1 kg
MP04800682 500 g
MP04800684 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP04800683 Supplier MP Biomedicals, Inc Supplier No. 04800683
Add to Cart
Add to Cart

Ethylenediamine Tetraacetic Acid, Electrophoresis Grade, >99.5%, White Powder

  • Ethylenediamine tetraacetic acid (EDTA) is a calcium chelator used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals.
  • For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used.
  • EDTA prevents platelet aggregation and is therefore the preferred anticoagulant for platelet counts.
  • It is an inhibitor of metalloproteases and metal-activated proteases.
  • It acts as a chelator of the zinc ion in the active site of metalloproteases, but EDTA can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases.
  • EDTA interferes with biological processes which are metal-dependent.
  • It is a buffer component.
  • Ethylenediamine Tetraacetic Acid is a polyamino carboxylic acid hexadentate ligand.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym EDTA, (Ethylenedinitrilo)tetraacetic acid, Edathamil, Ethylenedinitrilotetraacetic acid
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point 250°C (decomposition), 236°C to 246°C (literature)
Color White
Density ca. 860 kg/m3 (bulk density) (Lit.)
pH 2 to 3 (saturated solution)
Physical Form Powder
Quantity 1 kg
UN Number UN3077
Beilstein 1716295
Solubility Information Soluble in 3M NaOH (160mg/mL); 1 M aq soln: Clear, Colorless & Complete, No Black Specks, Haze Free.
RTECSNumber AH4025000
storageNote1 Store at Room Temperature(15-30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.