Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic acid tetrasodium salt hydrate, 98%

Catalog No. AAA173850I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA173850I 5000 g
AAA1738530 250 g
AAA173850B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA173850I Supplier Thermo Scientific Chemicals Supplier No. A173850I
Add to Cart

CAS: 194491-31-1 | C10H12N2Na4O8 | 380.17 g/mol

Ethylenediaminetetraacetic acid tetrasodium salt hydrate is used as an inhibitor of metal ligand proteases. It also acts as a chelator of zinc ion in the active site of metalloproteases. It is used as an anticoagulant and also used in the fluorometric determination of beryllium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid tetrasodium salt hydrate is used as an inhibitor of metal ligand proteases. It also acts as a chelator of zinc ion in the active site of metalloproteases. It is used as an anticoagulant and also used in the fluorometric determination of beryllium.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194491-31-1
Molecular Formula C10H12N2Na4O8
Molecular Weight (g/mol) 380.17
MDL Number MFCD00150025
InChI Key UEUXEKPTXMALOB-UHFFFAOYSA-J
Synonym edta tetrasodium salt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, tetrasodium 2-2-bis carboxylatomethyl amino ethyl carboxylatomethyl amino acetate hydrate, tetrasodium ion 4-edta hydrate, ethylenediaminetetraacetic acid, tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, bioultra kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, practical grade
PubChem CID 16211056
IUPAC Name tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;hydrate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point >300°C
Quantity 5000 g
Beilstein 3828865
Solubility Information Soluble in water.
Formula Weight 380.17 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Ethylenediaminetetraacetic acid tetrasodium salt hydrate
Hazard Category H302+H332-H318-H335
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
EINECSNumber 200-573-9
RTECSNumber AH5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.